Assay Details
| Gene Symbol: AKR1B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204629.2 | NP_989960.1 | 4 - 5 | 472 | 144 | |
| GenBank mRNA | AJ295030.1 | - | 4 - 5 | 472 | 144 | |
| BU297526.1 | - | 4 - 5 | 456 | 144 | ||
More Information
Set Membership: |
|

AKR1B10
Genomic Map