Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 27 - 28 | 3633 | 87 | |
| XM_003641367.4 | XP_003641415.1 | 29 - 30 | 4397 | 87 | ||
| XM_015286526.2 | XP_015142012.1 | 29 - 30 | 4623 | 87 | ||
| XM_015286527.2 | XP_015142013.1 | 30 - 31 | 4577 | 87 | ||
| XM_015286528.2 | XP_015142014.1 | 25 - 26 | 3963 | 87 | ||
| GenBank mRNA | AB046524.1 | - | 27 - 28 | 3633 | 87 | |
More Information
Set Membership: |
|

OVOA
Genomic Map