Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 32 - 33 | 4549 | 99 | |
| XM_003641367.4 | XP_003641415.1 | 34 - 35 | 5313 | 99 | ||
| XM_015286526.2 | XP_015142012.1 | 34 - 35 | 5539 | 99 | ||
| XM_015286527.2 | XP_015142013.1 | 35 - 36 | 5493 | 99 | ||
| XM_015286528.2 | XP_015142014.1 | 30 - 31 | 4879 | 99 | ||
| GenBank mRNA | AB046524.1 | - | 32 - 33 | 4549 | 99 | |
More Information
Set Membership: |
|

OVOA
Genomic Map