Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 34 - 35 | 4931 | 56 | |
| XM_003641367.4 | XP_003641415.1 | 36 - 37 | 5695 | 56 | ||
| XM_015286526.2 | XP_015142012.1 | 36 - 37 | 5921 | 56 | ||
| XM_015286527.2 | XP_015142013.1 | 37 - 38 | 5875 | 56 | ||
| XM_015286528.2 | XP_015142014.1 | 32 - 33 | 5261 | 56 | ||
| GenBank mRNA | AB046524.1 | - | 34 - 35 | 4931 | 56 | |
More Information
Set Membership: |
|

OVOA
Genomic Map