Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 6 - 7 | 920 | 110 | |
| XM_003641367.4 | XP_003641415.1 | 8 - 9 | 1684 | 110 | ||
| XM_015286526.2 | XP_015142012.1 | 8 - 9 | 1910 | 110 | ||
| XM_015286527.2 | XP_015142013.1 | 9 - 10 | 1864 | 110 | ||
| XM_015286528.2 | XP_015142014.1 | 4 - 5 | 1250 | 110 | ||
| GenBank mRNA | AB046524.1 | - | 6 - 7 | 920 | 110 | |
More Information
Set Membership: |
|

OVOA
Genomic Map