Assay Details
| Gene Symbol: SIK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204682.1 | NP_990013.1 | 12 - 13 | 2014 | 71 | |
| XM_025154772.1 | XP_025010540.1 | 15 - 16 | 6848 | 71 | ||
| XM_025154800.1 | XP_025010568.1 | 16 - 17 | 5932 | 71 | ||
| GenBank mRNA | AF219232.1 | - | 12 - 13 | 2014 | 71 | |
More Information
Set Membership: |
|

SIK1
Genomic Map