Assay Details
| Gene Symbol: HINT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204689.1 | NP_990020.1 | 1 - 2 | 144 | 91 | |
| GenBank mRNA | AB026675.1 | - | 1 - 2 | 144 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HINT2| Gene Symbol: HINT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204689.1 | NP_990020.1 | 1 - 2 | 144 | 91 | |
| GenBank mRNA | AB026675.1 | - | 1 - 2 | 144 | 91 | |
Set Membership: |
|