Assay Details
| Gene Symbol: RHOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204704.1 | NP_990035.1 | 2 - 3 | 202 | 85 | |
| XM_025154419.1 | XP_025010187.1 | 3 - 4 | 425 | 85 | ||
| XM_025154420.1 | XP_025010188.1 | 2 - 3 | 182 | 85 | ||
| GenBank mRNA | AF098513.1 | - | 2 - 3 | 231 | 85 | |
| AJ294705.1 | - | 2 - 3 | 202 | 85 | ||
| AJ720254.1 | - | 2 - 3 | 308 | 85 | ||
| U79757.1 | - | 1 - 2 | 154 | 85 | ||
More Information
Set Membership: |
|

RHOA
Genomic Map