Assay Details
| Gene Symbol: FGF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204714.1 | NP_990045.1 | 3 - 4 | 251 | 70 | |
| XM_015293588.2 | XP_015149074.1 | 4 - 5 | 633 | 70 | ||
| XM_015293589.2 | XP_015149075.1 | 3 - 4 | 502 | 70 | ||
| XM_015293590.2 | XP_015149076.1 | 2 - 3 | 336 | 70 | ||
| GenBank mRNA | AB030229.1 | - | 3 - 4 | 251 | 70 | |
| BX932351.1 | - | 3 - 4 | 366 | 70 | ||
More Information
Set Membership: |
|

FGF18
Genomic Map