Assay Details
| Gene Symbol: TSPAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204801.1 | NP_990132.1 | 7 - 8 | 763 | 64 | |
| GenBank mRNA | AF206661.1 | - | 7 - 8 | 763 | 64 | |
More Information
Set Membership: |
|

TSPAN18
Genomic Map