Assay Details
| Gene Symbol: LECT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204810.1 | NP_990141.1 | 4 - 5 | 464 | 94 | |
| GenBank mRNA | AF027380.1 | - | 4 - 5 | 463 | 94 | |
| AF138280.1 | - | 4 - 5 | 464 | 94 | ||
More Information
Set Membership: |
|

LECT1
Genomic Map