Assay Details
| Gene Symbol: CHRNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204812.2 | NP_990143.2 | 5 - 6 | 1245 | 94 | |
| XM_025144653.1 | XP_025000421.1 | 4 - 5 | 3114 | 94 | ||
| GenBank mRNA | X83739.2 | - | 94 | |||
More Information
Set Membership: |
|

CHRNB3
Genomic Map