Assay Details
| Gene Symbol: ELAVL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204830.1 | NP_990161.1 | 4 - 5 | 781 | 93 | |
| XM_015290948.2 | XP_015146434.1 | 4 - 5 | 628 | 93 | ||
| XM_015290949.2 | XP_015146435.2 | 4 - 5 | 612 | 93 | ||
| XM_015290950.2 | XP_015146436.1 | 4 - 5 | 628 | 93 | ||
| XM_015290951.2 | XP_015146437.1 | 4 - 5 | 625 | 93 | ||
| XM_015290952.2 | XP_015146438.1 | 4 - 5 | 628 | 93 | ||
| XM_015290953.2 | XP_015146439.1 | 4 - 5 | 556 | 93 | ||
| XM_015290954.2 | XP_015146440.1 | 4 - 5 | 553 | 93 | ||
| XM_015290955.2 | XP_015146441.1 | 4 - 5 | 666 | 93 | ||
| XM_015290956.2 | XP_015146442.1 | 4 - 5 | 617 | 93 | ||
| XM_015290957.2 | XP_015146443.1 | 4 - 5 | 674 | 93 | ||
| XM_015290959.2 | XP_015146445.1 | 4 - 5 | 518 | 93 | ||
| XM_025152752.1 | XP_025008520.1 | 4 - 5 | 605 | 93 | ||
| XM_025152754.1 | XP_025008522.1 | 4 - 5 | 576 | 93 | ||
| GenBank mRNA | AF176675.1 | - | 4 - 5 | 781 | 93 | |
More Information
Set Membership: |
|

ELAVL4
Genomic Map