Assay Details
| Gene Symbol: FOLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204834.1 | NP_990165.1 | 2 - 3 | 1111 | 84 | |
| XM_015280910.2 | XP_015136396.1 | 3 - 4 | 514 | 84 | ||
| GenBank mRNA | AF160960.1 | - | 2 - 3 | 1111 | 84 | |
More Information
Set Membership: |
|

FOLR1
Genomic Map