Assay Details
| Gene Symbol: NUMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204835.1 | NP_990166.1 | 7 - 8 | 905 | 63 | |
| XM_015286934.2 | XP_015142420.1 | 8 - 9 | 1025 | 63 | ||
| XM_015286935.2 | XP_015142421.1 | 8 - 9 | 1200 | 63 | ||
| XM_015286936.2 | XP_015142422.1 | 7 - 8 | 727 | 63 | ||
| XM_015286937.2 | XP_015142423.1 | 7 - 8 | 1020 | 63 | ||
| XM_015286938.2 | XP_015142424.1 | 8 - 9 | 980 | 63 | ||
| XM_015286939.2 | XP_015142425.1 | 7 - 8 | 947 | 63 | ||
| XM_015286940.2 | XP_015142426.1 | 6 - 7 | 649 | 63 | ||
| XM_015286941.2 | XP_015142427.1 | 8 - 9 | 1025 | 63 | ||
| XM_015286942.2 | XP_015142428.1 | 8 - 9 | 980 | 63 | ||
| XM_015286943.2 | XP_015142429.1 | 7 - 8 | 947 | 63 | ||
| XM_025150567.1 | XP_025006335.1 | 7 - 8 | 899 | 63 | ||
| XM_025150568.1 | XP_025006336.1 | 7 - 8 | 651 | 63 | ||
| GenBank mRNA | AF176086.1 | - | 7 - 8 | 905 | 63 | |
| JN638314.1 | - | 4 - 5 | 423 | 63 | ||
| JN638315.1 | - | 4 - 5 | 423 | 63 | ||
| KP756695.1 | - | 5 - 6 | 496 | 63 | ||
| KP756696.1 | - | 4 - 5 | 418 | 63 | ||
More Information
Set Membership: |
|

NUMB
Genomic Map