Assay Details
| Gene Symbol: LHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204889.1 | NP_990220.1 | 3 - 4 | 714 | 90 | |
| XM_015279326.2 | XP_015134812.2 | 3 - 4 | 1661 | 90 | ||
| GenBank mRNA | AB005882.1 | - | 3 - 4 | 714 | 90 | |
More Information
Set Membership: |
|

LHX2
Genomic Map