Assay Details
| Gene Symbol: SCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204890.1 | NP_990221.1 | 2 - 3 | 396 | 70 | |
| GenBank mRNA | X60465.1 | - | 396 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCD| Gene Symbol: SCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204890.1 | NP_990221.1 | 2 - 3 | 396 | 70 | |
| GenBank mRNA | X60465.1 | - | 396 | 70 | ||
Set Membership: |
|