Assay Details
| Gene Symbol: SERPINB10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204897.1 | NP_990228.1 | 3 - 4 | 324 | 99 | |
| XM_015282163.2 | XP_015137649.1 | 4 - 5 | 367 | 99 | ||
| XM_015282165.2 | XP_015137651.1 | 5 - 6 | 488 | 99 | ||
| GenBank mRNA | AF053401.1 | - | 3 - 4 | 324 | 99 | |
| BX932867.1 | - | 4 - 5 | 392 | 99 | ||
More Information
Set Membership: |
|

SERPINB10B
Genomic Map