Assay Details
| Gene Symbol: CNBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204907.1 | NP_990238.1 | 4 - 5 | 514 | 114 | |
| XM_015292957.2 | XP_015148443.1 | 4 - 5 | 575 | 114 | ||
| XM_015292958.1 | XP_015148444.1 | 6 - 7 | 1004 | 114 | ||
| XM_015292959.2 | XP_015148445.1 | 4 - 5 | 572 | 114 | ||
| XM_015292960.2 | XP_015148446.1 | 4 - 5 | 577 | 114 | ||
| XM_015292961.2 | XP_015148447.1 | 4 - 5 | 552 | 114 | ||
| XM_025154421.1 | XP_025010189.1 | 6 - 7 | 1359 | 114 | ||
| XM_025154423.1 | XP_025010191.1 | 6 - 7 | 690 | 114 | ||
| XM_025154424.1 | XP_025010192.1 | 5 - 6 | 754 | 114 | ||
| GenBank mRNA | AF004942.1 | - | 4 - 5 | 514 | 114 | |
| AF035676.1 | - | 4 - 5 | 524 | 114 | ||
More Information
Set Membership: |
|

CNBP
Genomic Map