Assay Details
| Gene Symbol: SH2D6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204908.1 | NP_990239.1 | 17 - 18 | 1227 | 65 | |
| XM_015288571.2 | XP_015144057.1 | 19 - 20 | 1333 | 65 | ||
| XM_015288572.2 | XP_015144058.1 | 18 - 19 | 1247 | 65 | ||
| XM_015288573.2 | XP_015144059.1 | 18 - 19 | 1267 | 65 | ||
| XM_015288574.2 | XP_015144060.1 | 16 - 17 | 1238 | 65 | ||
| XM_025151487.1 | XP_025007255.1 | 17 - 18 | 1206 | 65 | ||
| GenBank mRNA | AB015289.1 | - | 17 - 18 | 1206 | 65 | |
| AF089727.1 | - | 17 - 18 | 1227 | 65 | ||
More Information
Set Membership: |
|

SH2D6
Genomic Map