Assay Details
| Gene Symbol: MST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205213.1 | NP_990544.1 | 16 - 17 | 1825 | 92 | |
| GenBank mRNA | X84043.1 | - | 1825 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MST1| Gene Symbol: MST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205213.1 | NP_990544.1 | 16 - 17 | 1825 | 92 | |
| GenBank mRNA | X84043.1 | - | 1825 | 92 | ||
Set Membership: |
|