Assay Details
| Gene Symbol: WT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205216.1 | NP_990547.1 | 4 - 5 | 694 | 81 | |
| XM_015286134.2 | XP_015141620.1 | 4 - 5 | 81 | |||
| GenBank mRNA | AB033633.1 | - | 4 - 5 | 694 | 81 | |
| AB033634.1 | - | 4 - 5 | 694 | 81 | ||
| X85731.1 | - | 1063 | 81 | |||
More Information
Set Membership: |
|

WT1
Genomic Map