Assay Details
| Gene Symbol: CNGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205220.2 | NP_990551.1 | 2 - 3 | 338 | 91 | |
| XM_025149528.1 | XP_025005296.1 | 1 - 2 | 214 | 91 | ||
| GenBank mRNA | X89599.1 | - | 339 | 91 | ||
More Information
Set Membership: |
|

CNGA1
Genomic Map