Assay Details
| Gene Symbol: CNGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205221.1 | NP_990552.1 | 3 - 4 | 333 | 75 | |
| XM_015277691.2 | XP_015133177.1 | 3 - 4 | 649 | 75 | ||
| GenBank mRNA | X89598.1 | - | 333 | 75 | ||
More Information
Set Membership: |
|

CNGA3
Genomic Map