Assay Details
| Gene Symbol: RUNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205227.1 | NP_990558.1 | 5 - 6 | 878 | 61 | |
| XM_015299299.2 | XP_015154785.1 | 7 - 8 | 1071 | 61 | ||
| XM_015299305.2 | XP_015154791.1 | 6 - 7 | 879 | 61 | ||
| XM_025146751.1 | XP_025002519.1 | 7 - 8 | 1089 | 61 | ||
| XM_025146753.1 | XP_025002521.1 | 5 - 6 | 1264 | 61 | ||
| GenBank mRNA | Z36530.1 | - | 878 | 61 | ||
More Information
Set Membership: |
|

RUNX1
Genomic Map