Assay Details
| Gene Symbol: RBP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205238.1 | NP_990569.1 | 5 - 6 | 606 | 131 | |
| XM_025151488.1 | XP_025007256.1 | 5 - 6 | 599 | 131 | ||
| GenBank mRNA | X77960.1 | - | 606 | 131 | ||
More Information
Set Membership: |
|

RBP4A
Genomic Map