Assay Details
| Gene Symbol: HOXD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205249.1 | NP_990580.1 | 1 - 2 | 644 | 95 | |
| GenBank mRNA | JF317555.1 | - | 1 - 2 | 577 | 95 | |
| X62019.1 | - | 644 | 95 | |||
More Information
Set Membership: |
|

HOXD12
Genomic Map