Assay Details
Gene Symbol: SNRPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205250.1 | NP_990581.1 | 55 | 58 | ||
GenBank mRNA | BX934092.1 | - | 46 | 58 | ||
CR406596.1 | - | 113 | 58 | |||
L19064.1 | - | 55 | 58 | |||
X65702.1 | - | 55 | 58 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
