Assay Details
| Gene Symbol: SNRPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205250.1 | NP_990581.1 | 224 | 152 | ||
| GenBank mRNA | BX934092.1 | - | 571 | 152 | ||
| CR406596.1 | - | 628 | 152 | |||
| CR407342.1 | - | 718 | 152 | |||
| L19064.1 | - | 224 | 152 | |||
| X65702.1 | - | 224 | 152 | |||
More Information
Set Membership: |
|

SNRPE
Genomic Map