Assay Details
| Gene Symbol: HHEX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205252.1 | NP_990583.1 | 2 - 3 | 646 | 109 | |
| GenBank mRNA | X64711.1 | - | 646 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HHEX| Gene Symbol: HHEX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205252.1 | NP_990583.1 | 2 - 3 | 646 | 109 | |
| GenBank mRNA | X64711.1 | - | 646 | 109 | ||
Set Membership: |
|