Assay Details
| Gene Symbol: BMPR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205357.1 | NP_990688.1 | 5 - 6 | 672 | 69 | |
| XM_015287960.2 | XP_015143446.1 | 5 - 6 | 806 | 69 | ||
| XM_015287961.2 | XP_015143447.1 | 4 - 5 | 685 | 69 | ||
| GenBank mRNA | L49204.1 | - | 5 - 6 | 672 | 69 | |
More Information
Set Membership: |
|

BMPR1A
Genomic Map