Assay Details
| Gene Symbol: ACVR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205367.1 | NP_990698.1 | 3 - 4 | 1571 | 81 | |
| XM_015289792.2 | XP_015145278.1 | 3 - 4 | 648 | 81 | ||
| GenBank mRNA | D31899.1 | - | 3 - 4 | 372 | 81 | |
| U31222.1 | - | 3 - 4 | 1571 | 81 | ||
More Information
Set Membership: |
|

ACVR2A
Genomic Map