Assay Details
| Gene Symbol: NTRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205378.1 | NP_990709.1 | 11 - 12 | 1339 | 67 | |
| GenBank mRNA | U43396.1 | - | 11 - 12 | 1339 | 67 | |
More Information
Set Membership: |
|

NTRK1
Genomic Map