Assay Details
| Gene Symbol: INHBA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205396.1 | NP_990727.1 | 1 - 2 | 629 | 75 | |
| XM_015281320.2 | XP_015136806.1 | 2 - 3 | 607 | 75 | ||
| GenBank mRNA | U26946.1 | - | 1 - 2 | 629 | 75 | |
| U42377.1 | - | 2 - 3 | 571 | 75 | ||
More Information
Set Membership: |
|

INHBA
Genomic Map