Assay Details
| Gene Symbol: IRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205415.1 | NP_990746.1 | 6 - 7 | 667 | 105 | |
| XM_015294014.1 | XP_015149500.1 | 6 - 7 | 862 | 105 | ||
| XM_015294015.1 | XP_015149501.1 | 6 - 7 | 862 | 105 | ||
| GenBank mRNA | KC250010.1 | - | 5 - 6 | 547 | 105 | |
| L39766.1 | - | 6 - 7 | 667 | 105 | ||
More Information
Set Membership: |
|

IRF1
Genomic Map