Assay Details
| Gene Symbol: RBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205463.2 | NP_990794.1 | 2 - 3 | 201 | 76 | |
| XM_025152771.1 | XP_025008539.1 | 2 - 3 | 2906 | 76 | ||
| GenBank mRNA | BU267624.1 | - | 2 - 3 | 211 | 76 | |
| CR353084.1 | - | 2 - 3 | 201 | 76 | ||
| J03922.1 | - | 2 - 3 | 206 | 76 | ||
| X74247.1 | - | 182 | 76 | |||
More Information
Set Membership: |
|

RBP
Genomic Map