Assay Details
| Gene Symbol: RBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205463.2 | NP_990794.1 | 3 - 4 | 291 | 83 | |
| XM_025152771.1 | XP_025008539.1 | 3 - 4 | 2996 | 83 | ||
| GenBank mRNA | BU267624.1 | - | 3 - 4 | 301 | 83 | |
| CR353084.1 | - | 3 - 4 | 291 | 83 | ||
| J03922.1 | - | 3 - 4 | 296 | 83 | ||
| X74247.1 | - | 272 | 83 | |||
More Information
Set Membership: |
|

RBP
Genomic Map