Assay Details
| Gene Symbol: LECT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205478.2 | NP_990809.2 | 3 - 4 | 352 | 134 | |
| GenBank mRNA | CR388911.1 | - | 3 - 4 | 339 | 134 | |
| M29449.1 | - | 3 - 4 | 352 | 134 | ||
More Information
Set Membership: |
|

LECT2
Genomic Map