Assay Details
| Gene Symbol: HAPLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205482.1 | NP_990813.1 | 3 - 4 | 611 | 65 | |
| XM_015280597.2 | XP_015136083.1 | 4 - 5 | 733 | 65 | ||
| XM_015280598.2 | XP_015136084.1 | 3 - 4 | 1032 | 65 | ||
| XM_025144694.1 | XP_025000462.1 | 3 - 4 | 631 | 65 | ||
| GenBank mRNA | M13212.1 | - | 3 - 4 | 611 | 65 | |
More Information
Set Membership: |
|

HAPLN1
Genomic Map