Assay Details
| Gene Symbol: TIMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205487.2 | NP_990818.1 | 3 - 4 | 427 | 70 | |
| GenBank mRNA | CR386139.1 | - | 3 - 4 | 511 | 70 | |
| CR386544.1 | - | 3 - 4 | 492 | 70 | ||
| M94531.1 | - | 3 - 4 | 429 | 70 | ||
More Information
Set Membership: |
|

TIMP3
Genomic Map