Assay Details
| Gene Symbol: HSPA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205491.1 | NP_990822.1 | 8 - 9 | 1441 | 84 | |
| GenBank mRNA | M27260.1 | - | 8 - 9 | 1441 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSPA5| Gene Symbol: HSPA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205491.1 | NP_990822.1 | 8 - 9 | 1441 | 84 | |
| GenBank mRNA | M27260.1 | - | 8 - 9 | 1441 | 84 | |
Set Membership: |
|