Assay Details
| Gene Symbol: CST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205500.2 | NP_990831.2 | 1 - 2 | 290 | 62 | |
| GenBank mRNA | AJ395644.1 | - | 1 - 2 | 232 | 62 | |
| BU119189.1 | - | 1 - 2 | 225 | 62 | ||
| BU140104.1 | - | 1 - 2 | 228 | 62 | ||
| CN210879.1 | - | 1 - 2 | 279 | 62 | ||
| J05077.1 | - | 1 - 2 | 288 | 62 | ||
More Information
Set Membership: |
|

CST3
Genomic Map