Assay Details
| Gene Symbol: CD3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205512.1 | NP_990843.1 | 2 - 3 | 326 | 58 | |
| GenBank mRNA | AY574974.1 | - | 2 - 3 | 296 | 58 | |
| AY574975.1 | - | 2 - 3 | 299 | 58 | ||
| AY822036.1 | - | 2 - 3 | 299 | 58 | ||
| AY822037.1 | - | 2 - 3 | 299 | 58 | ||
| EU477524.1 | - | 2 - 3 | 296 | 58 | ||
| M59925.1 | - | 2 - 3 | 326 | 58 | ||
More Information
Set Membership: |
|

CD3D
Genomic Map