Assay Details
| Gene Symbol: CAPZA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205515.1 | NP_990846.1 | 742 | 92 | ||
| GenBank mRNA | AJ719691.1 | - | 432 | 92 | ||
| M25534.1 | - | 742 | 92 | |||
More Information
Set Membership: |
|

CAPZA1
Genomic Map