Assay Details
| Gene Symbol: PAM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155349.1 | XP_025011117.1 | 4 - 5 | 341 | 58 | |
| XM_025155350.1 | XP_025011118.1 | 4 - 5 | 374 | 58 | ||
| GenBank mRNA | BU237198.1 | - | 4 - 5 | 304 | 58 | |
| CN218549.1 | - | 4 - 5 | 330 | 58 | ||
More Information
Set Membership: |
|

PAM16
Genomic Map