Assay Details
| Gene Symbol: DTNBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006372.1 | NP_001006372.1 | 4 - 5 | 267 | 65 | |
| XM_015275875.2 | XP_015131361.1 | 4 - 5 | 240 | 65 | ||
| GenBank mRNA | AJ720064.1 | - | 4 - 5 | 267 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015275875.2 transcripts.
Set Membership: |
|

DTNBP1
Genomic Map