Assay Details
| Gene Symbol: TXNDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006374.1 | NP_001006374.1 | 2 - 3 | 403 | 62 | |
| XM_015275888.2 | XP_015131374.1 | 2 - 3 | 429 | 62 | ||
| XM_015275889.1 | XP_015131375.1 | 2 - 3 | 224 | 62 | ||
| GenBank mRNA | AJ720757.1 | - | 2 - 3 | 403 | 62 | |
More Information
Set Membership: |
|

TXNDC5
Genomic Map