Assay Details
| Gene Symbol: SSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006375.1 | NP_001006375.1 | 4 - 5 | 625 | 86 | |
| XM_015275890.2 | XP_015131376.1 | 8 - 9 | 2584 | 86 | ||
| XM_015275891.2 | XP_015131377.1 | 8 - 9 | 2584 | 86 | ||
| GenBank mRNA | AJ719521.1 | - | 4 - 5 | 625 | 86 | |
More Information
Set Membership: |
|

SSR1
Genomic Map