Assay Details
| Gene Symbol: DEPDC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006576.1 | NP_001006576.1 | 8 - 9 | 1097 | 70 | |
| GenBank mRNA | AJ719802.1 | - | 8 - 9 | 1134 | 70 | |
More Information
Set Membership: |
|

DEPDC1B
Genomic Map