Assay Details
| Gene Symbol: NOCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009928.1 | NP_001009928.1 | 2 - 3 | 424 | 66 | |
| XM_015276162.2 | XP_015131648.1 | 2 - 3 | 1265 | 66 | ||
| XM_025149538.1 | XP_025005306.1 | 2 - 3 | 435 | 66 | ||
| GenBank mRNA | AF503351.1 | - | 2 - 3 | 424 | 66 | |
More Information
Set Membership: |
|

NOCT
Genomic Map